Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031978.1 | NP_114184.1 | 14 | 1629 | ||
GenBank mRNA | AJ006340.1 | - | 14 | 1629 | ||
BC087654.1 | - | 14 | 1714 | |||
BC105883.1 | - | 14 | 1675 | |||
BC128734.1 | - | 14 | 1674 | |||
BC166420.1 | - | 14 | 1670 | |||
FQ218653.1 | - | 3 | 246 | |||
FQ234100.1 | - | 3 | 231 |