Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsfl1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031981.2 | NP_114187.1 | 2, 3 | 307 | ||
XM_006235254.2 | XP_006235316.1 | 2, 3 | 274 | |||
GenBank mRNA | AB002086.1 | - | 2, 3 | 307 | ||
BC072464.1 | - | 2, 3 | 204 | 7096463 | ||
FQ228953.1 | - | 2, 3 | 239 | |||
Y10769.1 | - | 2, 3 | 204 |