Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANK1 SIRNA Assays ›
Gene Symbol:
ANK1
Gene Name:
ankyrin 1
Gene Aliases:
ANK, SPH1, SPH2
Chromosome Location:
Chr.8: 41653225 - 41896762 on Build GRCh38
UniGene ID:
Hs.654438
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s1363
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANK1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000037.3
NP_000028.3
43
6240
NM_001142445.1
NP_001135917.1
4
1184
NM_001142446.1
NP_001135918.1
43
6426
NM_020475.2
NP_065208.2
42
6105
NM_020476.2
NP_065209.2
42
6180
NM_020477.2
NP_065210.2
42
5694
NM_020478.4
NP_065211.2
5
1244
NM_020480.4
NP_065213.2
3
1043
XM_005273476.4
XP_005273533.1
44
6538
XM_011544490.2
XP_011542792.1
45
6445
XM_011544491.2
XP_011542793.1
46
6504
XM_011544494.2
XP_011542796.1
44
6399
XM_011544495.2
XP_011542797.1
45
6458
XM_011544496.2
XP_011542798.1
45
6370
XM_011544500.2
XP_011542802.1
42
6282
XM_011544501.2
XP_011542803.1
43
6342
XM_011544502.2
XP_011542804.1
42
6207
XM_011544503.2
XP_011542805.1
45
5971
XM_011544504.2
XP_011542806.1
42
5796
XM_011544505.2
XP_011542807.1
43
5856
XM_017013319.1
XP_016868808.1
44
6420
XM_017013320.1
XP_016868809.1
45
6440
XM_017013321.1
XP_016868810.1
45
6389
XM_017013322.1
XP_016868811.1
44
6351
XM_017013323.1
XP_016868812.1
45
6365
XM_017013324.1
XP_016868813.1
43
6478
XM_017013325.1
XP_016868814.1
44
6290
XM_017013326.1
XP_016868815.1
44
6298
XM_017013327.1
XP_016868816.1
45
5958
XM_017013328.1
XP_016868817.1
44
5913
XM_017013329.1
XP_016868818.1
44
6050
GenBank mRNA
AA404587.1
-
1
119
AK129864.1
-
6
1061
X16609.1
-
42
6180
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
exocytosis
ER to Golgi vesicle-mediated transport
cytoskeleton organization
signal transduction
positive regulation of organelle organization
maintenance of epithelial cell apical/basal polarity
protein targeting to plasma membrane
structural molecule activity
structural constituent of cytoskeleton
protein binding
cytoskeletal adaptor activity
enzyme binding
spectrin binding
ATPase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-67fd5c5bdd-q6sk2:80/10.239.167.246:80.
git-commit: 7fb76fe7d0d673187a4c374cd16a9abd242c4866
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.3-offline