Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022208.1 | NP_071544.1 | 4 | 390 | ||
XM_006240410.2 | XP_006240472.1 | 4 | 665 | |||
GenBank mRNA | AF000943.1 | - | 4 | 390 |