Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032058.2 | NP_114447.2 | 3 | 445 | ||
GenBank mRNA | BC070506.1 | - | 3 | 445 | 7097686 | |
FQ232990.1 | - | 3 | 454 | |||
U31880.1 | - | 3 | 448 | |||
U83914.1 | - | 3 | 416 |