Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kalrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032062.2 | NP_114451.2 | 49 | 7268 | ||
GenBank mRNA | AF232669.1 | - | 48 | 7238 |