Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032085.1 | NP_114474.1 | 38, 39 | 2761 | ||
GenBank mRNA | BC087039.1 | - | 38, 39 | 2761 | 7097081 | |
X70369.1 | - | 3, 4 | 177 |