Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SURF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001280787.1 | NP_001267716.1 | 6 | 669 | ||
NM_003172.3 | NP_003163.1 | 7 | 721 | |||
XM_011518942.1 | XP_011517244.1 | 6 | 771 | |||
GenBank mRNA | AK291122.1 | - | 7 | 704 | ||
BC028314.1 | - | 7 | 709 | 5190921 | ||
BC071658.1 | - | 7 | 726 | 5539506 | ||
KU178341.1 | - | 7 | 689 | |||
KU178342.1 | - | 6 | 637 | |||
KU178343.1 | - | 5 | 503 | |||
Z35093.1 | - | 7 | 703 |