Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ikbkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053355.2 | NP_445807.2 | 22 | 2331 | ||
GenBank mRNA | AF115282.1 | - | 21 | 2245 |