Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053359.2 | NP_445811.1 | 2, 3 | 185 | ||
GenBank mRNA | AF127137.1 | - | 2, 3 | 121 | ||
AF177671.1 | - | 2, 3 | 200 | |||
BC058458.1 | - | 2, 3 | 209 | 6919265 | ||
FQ220104.1 | - | 2, 3 | 186 |