Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3kbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053360.2 | NP_445812.1 | 8 | 1089 | ||
XM_006256924.2 | XP_006256986.2 | 8 | 1031 | |||
XM_006256929.2 | XP_006256991.2 | 6 | 857 | |||
XM_006256930.2 | XP_006256992.1 | 3 | 450 | |||
XM_006256931.1 | XP_006256993.1 | 10 | 1382 | |||
GenBank mRNA | AF131867.1 | - | 6 | 645 | ||
AF230519.1 | - | 8 | 901 | |||
AF230520.1 | - | 8 | 755 | |||
AF255886.1 | - | 4 | 326 | |||
AF255887.1 | - | 3 | 220 | |||
AF255888.1 | - | 8 | 1007 | |||
BC070877.1 | - | 8 | 1089 | 7096160 |