Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168670.1 | NP_001162141.1 | 2 | 616 | ||
GenBank mRNA | AF131912.1 | - | 1 | 217 |