Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pglyrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053373.1 | NP_445825.1 | 2 | 294 | ||
GenBank mRNA | AF154114.1 | - | 2 | 294 |