Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053379.3 | NP_445831.3 | 5, 6 | 1006 | ||
XM_006257382.2 | XP_006257444.1 | 5, 6 | 1131 | |||
XM_006257385.2 | XP_006257447.1 | 5, 6 | 1224 | |||
GenBank mRNA | AF155959.2 | - | 4, 5 | 940 |