Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SURF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278942.1 | NP_001265871.1 | 5 | 725 | ||
NM_006753.5 | NP_006744.2 | 5 | 726 | |||
NR_103874.1 | 5 | 673 | ||||
GenBank mRNA | AK054884.1 | - | 5 | 723 | ||
BC003001.1 | - | 5 | 653 | |||
BC006197.2 | - | 5 | 653 | 2820773 | ||
BC014878.1 | - | 5 | 688 | 4343122 | ||
BI820270.1 | - | 5 | 668 | |||
BM451240.1 | - | 5 | 725 |