Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Puf60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191880.1 | NP_001178809.1 | 12 | 1422 | ||
XM_006241894.2 | XP_006241956.1 | 12 | 1558 | |||
XM_006241895.2 | XP_006241957.1 | 11 | 1507 | |||
XM_006241896.2 | XP_006241958.1 | 11 | 1504 | |||
XM_006241897.2 | XP_006241959.1 | 11 | 1456 | |||
XM_006241898.2 | XP_006241960.1 | 10 | 1405 | |||
XM_006241899.2 | XP_006241961.1 | 12 | 1851 | |||
GenBank mRNA | AF165892.1 | - | 11 | 1411 |