Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfra3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053398.1 | NP_445850.1 | 6 | 979 | ||
GenBank mRNA | AF184920.1 | - | 4 | 519 | ||
BC079378.1 | - | 6 | 979 | 7110848 |