Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053402.2 | NP_445854.1 | 2 | 173 | ||
GenBank mRNA | AF188608.1 | - | 2 | 174 | ||
BC098752.1 | - | 2 | 194 | 7369699 |