Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297438.1 | NP_001284367.1 | 2 | 450 | ||
NM_014231.4 | NP_055046.1 | 2 | 450 | |||
NM_016830.3 | NP_058439.1 | 2 | 450 | |||
NM_199245.2 | NP_954740.1 | 2 | 450 | |||
GenBank mRNA | AF060538.1 | - | 2 | 212 | ||
AK289498.1 | - | 2 | 221 | |||
AK296294.1 | - | 2 | 228 | |||
AK303137.1 | - | 2 | 228 | |||
BC023286.1 | - | 2 | 216 | 4943992 | ||
BE246448.1 | - | 2 | 228 | |||
BI838752.1 | - | 2 | 245 | |||
BQ717992.1 | - | 2 | 209 | |||
CR542214.1 | - | 1 | 82 | |||
CR542231.1 | - | 1 | 82 | |||
CR749485.1 | - | 2 | 451 | |||
CR989458.1 | - | 2 | 204 | |||
Z48924.1 | - | 1 | 82 |