Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053428.1 | NP_445880.1 | 4 | 504 | ||
XM_006257592.1 | XP_006257654.1 | 4 | 772 | |||
XM_008773572.1 | XP_008771794.1 | 5 | 680 | |||
XM_008773573.1 | XP_008771795.1 | 5 | 742 | |||
GenBank mRNA | AF271786.1 | - | 4 | 504 |