Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rims1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052829.1 | NP_439894.1 | 21 | 3650 | ||
XM_008766936.1 | XP_008765158.1 | 21 | 3650 | |||
GenBank mRNA | AF007836.1 | - | 21 | 3650 | ||
AF199333.1 | - | 21 | 3650 |