Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otof | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276720.1 | NP_001263649.1 | 38 | 4902 | ||
XM_006239833.2 | XP_006239895.1 | 39 | 4950 | |||
XM_008764534.1 | XP_008762756.1 | 38 | 4905 | |||
XM_008764535.1 | XP_008762757.1 | 38 | 4965 | |||
GenBank mRNA | AF315944.1 | - | 1 | 121 |