Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cetn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229519.1 | XP_006229581.1 | 2 | 275 | ||
XM_215222.7 | XP_215222.5 | 2 | 411 | |||
GenBank mRNA | AF334108.1 | - | 1 | 19 | ||
FQ219097.1 | - | 2 | 169 |