Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053464.2 | NP_445916.1 | 4 | 579 | ||
GenBank mRNA | AF337636.1 | - | 4 | 582 | ||
BC128733.1 | - | 4 | 625 | 8366755 |
Gene Symbol: LOC100912604 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749458.2 | XP_003749506.1 | 4 | 576 |