Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Socs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_058208.1 | NP_478115.1 | 2 | 433 | ||
GenBank mRNA | AF075382.1 | - | 2 | 433 |