Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spa17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053482.1 | NP_445934.1 | 1 | 103 | ||
XM_006242812.2 | XP_006242874.1 | 2 | 279 | |||
GenBank mRNA | AJ131888.1 | - | 1 | 103 |