Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnga3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053495.1 | NP_445947.1 | 5 | 1127 | ||
XM_008767042.1 | XP_008765264.1 | 7 | 1651 | |||
XM_008767043.1 | XP_008765265.1 | 6 | 1537 | |||
XM_008767044.1 | XP_008765266.1 | 6 | 1534 | |||
GenBank mRNA | AB002801.1 | - | 5 | 1127 | ||
AF031943.1 | - | 1 | 328 | |||
AJ272428.1 | - | 7 | 1414 | |||
AJ272429.1 | - | 6 | 1300 |