Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191856.2 | NP_001178785.1 | 2 | 898 | ||
GenBank mRNA | AF100787.1 | - | 2 | 143 |