Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053513.1 | NP_445965.1 | 9 | 1106 | ||
XM_008768127.1 | XP_008766349.1 | 8 | 2511 | |||
XM_008768128.1 | XP_008766350.1 | 9 | 2180 | |||
XM_008768129.1 | XP_008766351.1 | 9 | 1192 | |||
XM_008768130.1 | XP_008766352.1 | 8 | 1037 | |||
GenBank mRNA | FQ211962.1 | - | 9 | 1291 |