Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053520.2 | NP_445972.2 | 2 | 409 | ||
XM_006252347.2 | XP_006252409.1 | 3 | 1668 | |||
XM_006252348.2 | XP_006252410.1 | 2 | 406 | |||
XM_006252349.2 | XP_006252411.1 | 4 | 1834 | |||
XM_006252351.2 | XP_006252413.1 | 3 | 912 | |||
XM_006252352.2 | XP_006252414.1 | 3 | 872 | |||
XM_006252354.2 | XP_006252416.1 | 3 | 1668 | |||
XM_008770898.1 | XP_008769120.1 | 3 | 3172 | |||
XM_008770899.1 | XP_008769121.1 | 3 | 868 | |||
XM_008770900.1 | XP_008769122.1 | 3 | 424 | |||
XM_008770901.1 | XP_008769123.1 | 3 | 3172 | |||
XM_008770902.1 | XP_008769124.1 | 3 | 3172 | |||
XM_008770903.1 | XP_008769125.1 | 3 | 3172 | |||
GenBank mRNA | AB030216.1 | - | 2 | 178 | ||
AB030217.1 | - | 2 | 178 | |||
AB046531.1 | - | 2 | 188 | |||
CB782606.1 | - | 3 | 464 | |||
FQ221456.1 | - | 4 | 977 | |||
FQ233920.1 | - | 3 | 483 |