Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020783.3 | NP_065834.1 | 2 | 1048 | ||
GenBank mRNA | AB037763.1 | - | 2 | 865 | ||
AF299075.1 | - | 2 | 772 | |||
AK291154.1 | - | 2 | 888 | |||
AK293791.1 | - | 2 | 834 | |||
AK299593.1 | - | 3 | 291 | |||
AK299652.1 | - | 3 | 544 | |||
BC036538.2 | - | 2 | 777 | 5260337 |