Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Twist1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053530.2 | NP_445982.1 | 2 | 891 | ||
GenBank mRNA | BC166412.1 | - | 2 | 1021 | 9033082 |