Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297774.1 | NP_001284703.1 | 5 | 1041 | ||
NM_003180.2 | NP_003171.2 | 7 | 1096 | |||
XM_006723339.3 | XP_006723402.1 | 7 | 931 | |||
XM_006723340.3 | XP_006723403.1 | 7 | 1182 | |||
XM_006723341.3 | XP_006723404.1 | 5 | 1515 | |||
XM_017027175.1 | XP_016882664.1 | 7 | 993 | |||
XM_017027176.1 | XP_016882665.1 | 7 | 984 | |||
GenBank mRNA | AK125182.1 | - | 6 | 1884 | ||
AK295337.1 | - | 5 | 988 | |||
AK313067.1 | - | 7 | 885 | |||
BC029962.1 | - | 7 | 1024 | |||
BC033683.1 | - | 7 | 900 | |||
BC046157.1 | - | 7 | 1022 | 5768093 | ||
DQ090951.1 | - | 6 | 812 |