Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134452.1 | NP_604447.1 | 4 | 639 | ||
XM_006233873.2 | XP_006233935.1 | 4 | 1045 | |||
GenBank mRNA | AF272662.1 | - | 4 | 639 |