Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053532.1 | NP_445984.1 | 3 | 225 | ||
GenBank mRNA | AF285103.1 | - | 3 | 225 | ||
BC060551.1 | - | 3 | 235 | 6922407 | ||
FQ210403.1 | - | 2 | 118 | |||
FQ212413.1 | - | 2 | 118 | |||
FQ212825.1 | - | 2 | 118 | |||
FQ213190.1 | - | 2 | 118 | |||
FQ213755.1 | - | 2 | 118 | |||
FQ215265.1 | - | 2 | 118 | |||
FQ216419.1 | - | 2 | 118 | |||
FQ218632.1 | - | 2 | 118 | |||
FQ219250.1 | - | 2 | 118 | |||
FQ219494.1 | - | 2 | 118 | |||
FQ219779.1 | - | 2 | 118 | |||
FQ220023.1 | - | 2 | 118 | |||
FQ224099.1 | - | 3 | 231 | |||
FQ229108.1 | - | 2 | 118 | |||
FQ229165.1 | - | 2 | 118 | |||
FQ229238.1 | - | 2 | 118 | |||
FQ229410.1 | - | 2 | 118 | |||
FQ229570.1 | - | 2 | 118 | |||
FQ229620.1 | - | 2 | 118 | |||
FQ229735.1 | - | 2 | 118 | |||
FQ229750.1 | - | 2 | 118 | |||
FQ230042.1 | - | 2 | 118 | |||
FQ232691.1 | - | 2 | 118 |