Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: T | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270484.1 | NP_001257413.1 | 5 | 1047 | ||
NM_003181.3 | NP_003172.1 | 5 | 1140 | |||
XM_011536080.2 | XP_011534382.1 | 4 | 907 | |||
GenBank mRNA | AJ001699.1 | - | 4 | 780 | ||
BC039877.1 | - | 4 | 907 | |||
BC098425.1 | - | 5 | 1020 | 6424435 |