Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chi3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053560.1 | NP_446012.1 | 3 | 276 | ||
XM_006249892.2 | XP_006249954.2 | 3 | 330 | |||
XM_008769574.1 | XP_008767796.1 | 3 | 297 | |||
GenBank mRNA | AF062038.1 | - | 1 | 157 | ||
BC091365.1 | - | 3 | 276 | 7319171 | ||
EF467168.1 | - | 1 | 157 |