Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx39a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053563.2 | NP_446015.2 | 6 | 935 | ||
XM_006255314.2 | XP_006255376.1 | 6 | 835 | |||
GenBank mRNA | AF063447.1 | - | 6 | 731 | ||
BC086328.1 | - | 6 | 935 | 7132938 | ||
FQ231639.1 | - | 6 | 841 |