Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vegp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053574.1 | NP_446026.1 | 4, 5 | 456 | ||
GenBank mRNA | X74806.1 | - | 4, 5 | 456 |
Gene Symbol: LOC100911507 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233718.2 | XP_006233780.1 | 169 |