Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gosr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053584.2 | NP_446036.1 | 6 | 480 | ||
XM_006246914.1 | XP_006246976.1 | 6 | 548 | |||
XM_008768132.1 | XP_008766354.1 | 5 | 351 | |||
GenBank mRNA | BC126068.1 | - | 6 | 484 | 7387185 |