Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fez2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053600.2 | NP_446052.2 | 6 | 934 | ||
GenBank mRNA | AB076183.1 | - | 6 | 934 | ||
AF120109.1 | - | 1 | 27 | |||
CB725415.1 | - | 2 | 176 | |||
U64689.1 | - | 6 | 760 |