Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sbk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147135.2 | NP_671476.1 | 2, 3 | 718 | ||
XM_008759853.1 | XP_008758075.1 | 2, 3 | 725 | |||
XM_008759854.1 | XP_008758076.1 | 3, 4 | 1053 | |||
GenBank mRNA | AB010154.1 | - | 2, 3 | 430 | ||
BC131843.1 | - | 2, 3 | 718 | 8363395 |
Gene Symbol: LOC103690016 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758862.1 | XP_008757084.1 | 2, 3 | 766 | ||
XM_008758863.1 | XP_008757085.1 | 2, 3 | 723 | |||
XM_008758864.1 | XP_008757086.1 | 3, 4 | 1027 |