Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053616.1 | NP_446068.1 | 10 | 1471 | ||
GenBank mRNA | BC072524.1 | - | 10 | 1465 | 7097375 | |
U93692.1 | - | 10 | 1471 |