Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc42bpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053620.3 | NP_446072.2 | 28, 29 | 3836 | ||
XM_006240562.2 | XP_006240624.1 | 29, 30 | 4036 | |||
GenBank mRNA | AF021936.1 | - | 28, 29 | 3796 | ||
AY277590.1 | - | 28, 29 | 3836 |