Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cml5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080884.1 | NP_543160.1 | 2 | 1039 | ||
XM_006236720.2 | XP_006236782.1 | 2 | 918 | |||
XM_006236721.2 | XP_006236783.1 | 2 | 2447 | |||
XM_006236722.2 | XP_006236784.1 | 2 | 944 | |||
XM_006236723.1 | XP_006236785.1 | 1 | 683 | |||
XM_006236725.2 | XP_006236787.1 | 2 | 702 | |||
XM_008763044.1 | XP_008761266.1 | 3 | 1006 | |||
GenBank mRNA | AF187100.1 | - | 1 | 371 | ||
BC158648.1 | - | 2 | 1039 | 8367103 |