Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Megf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053628.2 | NP_446080.1 | 23 | 4377 | ||
XM_006228364.2 | XP_006228426.1 | 23 | 4723 |