Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053635.2 | NP_446087.1 | 16 | 1949 | ||
GenBank mRNA | AB037898.1 | - | 16 | 1823 | ||
AB049189.1 | - | 16 | 1949 | |||
BC097271.1 | - | 16 | 1854 | 7445508 |