Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stxbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053637.2 | NP_446089.1 | 9 | 898 | ||
GenBank mRNA | AB046544.1 | - | 9 | 899 | ||
BC098660.1 | - | 9 | 826 | 7386801 |