Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdlim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053650.1 | NP_446102.1 | 2 | 292 | ||
XM_006253119.2 | XP_006253181.1 | 2 | 327 | |||
XM_006253120.2 | XP_006253182.1 | 2 | 325 | |||
GenBank mRNA | AF002281.1 | - | 2 | 310 | ||
BC081703.1 | - | 2 | 284 | 7193873 | ||
FQ215471.1 | - | 2 | 307 |