Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053655.3 | NP_446107.2 | 9 | 1219 | ||
XM_006248715.2 | XP_006248777.1 | 10 | 1269 | |||
XM_006248716.2 | XP_006248778.1 | 10 | 1251 | |||
XM_006248717.2 | XP_006248779.1 | 10 | 1268 | |||
XM_006248718.2 | XP_006248780.1 | 9 | 1229 | |||
XM_006248719.2 | XP_006248781.1 | 9 | 1213 | |||
XM_006248720.2 | XP_006248782.1 | 10 | 1268 | |||
XM_006248721.2 | XP_006248783.1 | 10 | 1268 | |||
XM_006248722.2 | XP_006248784.1 | 9 | 1229 | |||
XM_006248723.2 | XP_006248785.1 | 10 | 1250 | |||
XM_006248724.2 | XP_006248786.1 | 9 | 1213 | |||
XM_008768840.1 | XP_008767062.1 | 10 | 1269 | |||
XM_008768841.1 | XP_008767063.1 | 10 | 1251 | |||
XM_008768842.1 | XP_008767064.1 | 9 | 1230 | |||
XM_008768843.1 | XP_008767065.1 | 10 | 1251 | |||
XM_008768844.1 | XP_008767066.1 | 9 | 1213 | |||
XM_008768845.1 | XP_008767067.1 | 10 | 1268 | |||
XM_008768846.1 | XP_008767068.1 | 9 | 1213 | |||
XM_008768847.1 | XP_008767069.1 | 10 | 1268 | |||
GenBank mRNA | BC085843.1 | - | 9 | 1219 | 7135909 |